Assay Details
| Gene Symbol: Gm3364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090251.1 | - | 1 - 2 | 873 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 36017 |
Gm36017 | -273.2kb | Sense | Chr. 5 - 44412732 - 44427652 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930431F12 gene | 4930431F12Rik | 260.1kb | Anti-sense | Chr. 5 - 44966201 - 44978805 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm3364
Genomic Map