Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 15984 | Gm15984 | OTTMUSG00000028614 | -21.3kb | Sense | Chr. 5 - 74702454 - 74728600 on NCBI Build GRCm38 |
| Downstream | predicted gene 6116 | Gm6116 | EG619945 | 195.8kb | Sense | Chr. 5 - 74949194 - 74966368 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map