Assay Details
| Gene Symbol: Dreh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105051.1 | 1 - 2 | 237 | 84 | ||
| GenBank mRNA | AK050349.1 | - | 1 - 2 | 238 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
mannosidase 2, alpha 1 |
Man2a1 |
Mana-2, Mana2, Map-2 |
-12.0kb | Sense | Chr. 17 - 64601649 - 64755110 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930583I09 gene | 4930583I09Rik | 64.6kb | Anti-sense | Chr. 17 - 64832523 - 64836107 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Dreh
Genomic Map