Assay Details
| Gene Symbol: 4930415C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015143.1 | - | 1 - 2 | 264 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 34245 |
Gm34245 | Sense | Chr. 13 - 59070788 - 59082920 on NCBI Build GRCm38 | ||
| Downstream | ATP/GTP binding protein 1 | Agtpbp1 |
1700020N17Rik, 2310001G17Rik, 2900054O13Rik, 4930445M19Rik, 5730402G09Rik, CCP1, Nna1, nmf243, pcd |
365.4kb | Sense | Chr. 13 - 59449534 - 59557364 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930415C11Rik
Genomic Map