Assay Details
| Gene Symbol: 2410080I02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019121.1 | - | 1 - 2 | 56 | 66 | |
| AK019124.1 | - | 1 - 2 | 56 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA C330002G04 gene | C330002G04Rik | -59.0kb | Anti-sense | Chr. 19 - 22996890 - 23075906 on NCBI Build GRCm38 | |
| Downstream | Kruppel-like factor 9 | Klf9 |
2310051E17Rik, AA589643, BTEB-1, Bteb1, ENSMUSG00000055440, Gm9971 |
5.5kb | Sense | Chr. 19 - 23141226 - 23166911 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

2410080I02Rik
Genomic Map