Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300012936.mm.1 |
NM_001025261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012936.mm.1 |
NM_001025261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012936.mm.1 |
NM_001025262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012936.mm.1 |
NM_001025262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012936.mm.1 |
NM_001025263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012936.mm.1 |
NM_001025263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012936.mm.1 |
NM_001025264.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012936.mm.1 |
NM_001025264.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012936.mm.1 |
NM_009412.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012936.mm.1 |
NM_009412.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012936.mm.1 |
TC0300001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012937.mm.1 |
NM_001025261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012937.mm.1 |
NM_001025261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012937.mm.1 |
NM_001025262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012937.mm.1 |
NM_001025262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012937.mm.1 |
NM_001025263.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012937.mm.1 |
NM_001025263.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012937.mm.1 |
NM_001025264.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012937.mm.1 |
NM_001025264.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012937.mm.1 |
NM_009412.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012937.mm.1 |
NM_009412.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012937.mm.1 |
TC0300001624.mm.1 |
MTA TC |