Assay Details
| Gene Symbol: F630040K05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080425.1 | - | 4 - 5 | 560 | 119 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627994 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54335 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 408906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588002 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600812 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128359 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672785 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56259 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645716 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649951 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850248 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 37403 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875294 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377584 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397386 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673377 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350821 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878708 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8395 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35668 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48058 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140993 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133077 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174737 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133077 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140993 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174737 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03323530 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103059 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133077 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140993 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13428378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13542176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13570914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13446839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13190359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13425163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13350248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13570919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13376475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13521204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13376547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13220362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13191614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13542169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13425164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13220368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13220366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13570873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13373732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13118480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13157982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13510644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13350249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13498820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13073249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13376530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13570872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13446838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13425161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13521194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13421541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13474208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13474212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13289591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13420009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13356176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13253180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13336901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13428382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13428379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13289589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13570885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13149844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31202849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13510647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31203015 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103520279 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103520279 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2582821 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103520279 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103520279 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2582820 | - | - |

F630040K05Rik
Genomic Map