Assay Details
| Gene Symbol: Zbtb33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079513.1 | NP_001072981.1 | 1 - 2 | 116 | 90 | |
| GenBank mRNA | AK087417.1 | - | 1 - 2 | 98 | 90 | |
| BY200826.1 | - | 1 - 2 | 117 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087417.1, NM_001079513.1 transcripts.
Set Membership: |
|

Zbtb33
Genomic Map