Assay Details
| Gene Symbol: Nudt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291044.1 | NP_001277973.1 | 3 - 4 | 520 | 114 | |
| GenBank mRNA | AF453427.1 | - | 3 - 4 | 629 | 114 | |
| AF453428.1 | - | 3 - 4 | 506 | 114 | ||
| AK145299.1 | - | 3 - 4 | 512 | 114 | ||
| BC146000.1 | - | 3 - 4 | 514 | 40131303 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF453427.1 transcripts.
Set Membership: |
|

Nudt6
Genomic Map