Assay Details
| Gene Symbol: Atg4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174874.3 | NP_777363.1 | 3 - 4 | 196 | 152 | |
| GenBank mRNA | AJ504653.1 | - | 2 - 3 | 188 | 152 | |
| AK010949.1 | - | 3 - 4 | 324 | 152 | ||
| AK028712.1 | - | 3 - 4 | 207 | 152 | ||
| AK031570.1 | - | 3 - 4 | 197 | 152 | ||
| AK075796.1 | - | 3 - 4 | 207 | 152 | ||
| AK088811.1 | - | 3 - 4 | 269 | 152 | ||
| AK129242.1 | - | 2 - 3 | 184 | 152 | ||
| AK144239.1 | - | 3 - 4 | 202 | 152 | ||
| AK188147.1 | - | 1 - 2 | 45 | 152 | ||
| BC027184.1 | - | 3 - 4 | 222 | 4237950 | 152 | |
More Information
Set Membership: |
|

Atg4b
Genomic Map