Assay Details
| Gene Symbol: Gskip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178613.3 | NP_848728.2 | 1 - 2 | 110 | 154 | |
| GenBank mRNA | AK017049.1 | - | 634 | 154 | ||
| AK020785.1 | - | 1 - 2 | 48 | 154 | ||
| AK042664.1 | - | 1 - 2 | 71 | 154 | ||
| AK079775.1 | - | 1 - 2 | 56 | 154 | ||
| AK168962.1 | - | 1 - 2 | 74 | 154 | ||
| AK171097.1 | - | 1 - 2 | 87 | 154 | ||
| BC031494.1 | - | 1 - 2 | 78 | 3156862 | 154 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK042664.1, AK079775.1, AK168962.1, AK171097.1, BC031494.1, NM_178613.3 transcripts.
Set Membership: |
|

Gskip
Genomic Map