Assay Details
| Gene Symbol: Ptbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077363.2 | NP_001070831.1 | 1 - 2 | 291 | 122 | |
| NM_001283013.1 | NP_001269942.1 | 1 - 2 | 291 | 122 | ||
| NM_008956.3 | NP_032982.2 | 1 - 2 | 291 | 122 | ||
| GenBank mRNA | AK037607.1 | - | 1 - 2 | 91 | 122 | |
| AK082600.1 | - | 1 - 2 | 95 | 122 | ||
| AK150840.1 | - | 1 - 2 | 95 | 122 | ||
| AK151292.1 | - | 1 - 2 | 119 | 122 | ||
| AK153561.1 | - | 1 - 2 | 119 | 122 | ||
| AK163274.1 | - | 1 - 2 | 116 | 122 | ||
| AK168392.1 | - | 1 - 2 | 92 | 122 | ||
| AK172712.1 | - | 1 - 2 | 91 | 122 | ||
| BC007472.1 | - | 1 - 2 | 108 | 3601527 | 122 | |
| BY167466.1 | - | 1 - 2 | 119 | 122 | ||
| CJ075278.1 | - | 1 - 2 | 297 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK082600.1, AK150840.1, AK151292.1, AK153561.1, AK163274.1, AK172712.1, BC007472.1, NM_001077363.2, NM_001283013.1, NM_008956.3 transcripts.
Set Membership: |
|

Ptbp1
Genomic Map