Assay Details
| Gene Symbol: Hnrnpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170983.1 | NP_001164454.1 | 6 - 7 | 732 | 175 | |
| GenBank mRNA | AK019958.1 | - | 6 - 7 | 1907 | 175 | |
| AK088678.1 | - | 6 - 7 | 743 | 175 | ||
| AK166590.1 | - | 6 - 7 | 710 | 175 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnpc
Genomic Map