Assay Details
| Gene Symbol: Apol9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168660.1 | NP_001162131.1 | 1 - 2 | 127 | 165 | |
| GenBank mRNA | AK009386.1 | - | 1 - 2 | 128 | 165 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001168660.1 transcripts.
Set Membership: |
|

Apol9b
Genomic Map