Assay Details
| Gene Symbol: 4933431E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015459.1 | 3 | 2583 | 129 | ||
| GenBank mRNA | AK030251.1 | - | 4 | 2205 | 129 | |
| AK036227.1 | - | 3 | 2156 | 129 | ||
| AK147824.1 | - | 4 | 2180 | 129 | ||
| BC060136.1 | - | 3 | 2583 | 129 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177858.1 | - | 2156 | Chr. - - [-] on Build | 129 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933431E20Rik
Genomic Map