Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA let7a-1 | Mirlet7a-1 |
Let-7a, Mirnlet7a, Mirnlet7a-1, let-7a-1 |
-4.4kb | Sense | Chr. 13 - 48538179 - 48538272 on NCBI Build GRCm38 |
| Downstream | protein tyrosine phosphatase domain containing 1 | Ptpdc1 |
AI843923, AW456874, Naa-1, Ptpcd1 |
34.5kb | Sense | Chr. 13 - 48577869 - 48625672 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map