Assay Details
| Gene Symbol: Jpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137473.1 | - | 3 | 490 | 116 | |
| AK148110.1 | - | 3 | 509 | 116 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
X (inactive)-specific transcript, opposite strand |
Tsix | -17.9kb | Sense | Chr. X - 103431517 - 103484957 on NCBI Build GRCm38 | |
| Downstream | polycomb group ring finger 6 pseudogene | Gm9785 | ENSMUSG00000043661 | 22.0kb | Anti-sense | Chr. X - 103525319 - 103525847 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Jpx
Genomic Map