Assay Details
| Gene Symbol: Ptrh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098810.2 | NP_001092280.1 | ||||
| NM_175004.2 | NP_778169.1 | |||||
| GenBank mRNA | AK028555.1 | - | ||||
| AK031620.1 | - | |||||
| AK038888.1 | - | |||||
| AK086996.1 | - | |||||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27990329 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022297 | AK038888.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022297 | AK086996.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022297 | AK028555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022297 | AK031620.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022297 | NM_001098810.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468795 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335657 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446290 | AK080710.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446290 | BC048943.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446290 | XM_127170.7 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446290 | XM_902085.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446290 | XM_915517.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446290 | XM_923785.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880634 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508646 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45874 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12364 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167485 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022297 | AK038888.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022297 | AK086996.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022297 | AK028555.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022297 | AK031620.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022297 | NM_001098810.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174555 | AK038888.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174555 | AK086996.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174555 | AK028555.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174555 | AK031620.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174555 | NM_001098810.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P123315 | AK038888.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P123315 | AK086996.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P123315 | BC026947.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P123315 | AK028555.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P123315 | AK031620.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27990358 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167485 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07429257 | - | - |
| Whole Mouse Genome G4122A | A_52_P123315 | AK038888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123315 | AK086996.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123315 | BC026947.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123315 | AK028555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123315 | AK031620.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174555 | AK038888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174555 | AK086996.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174555 | AK028555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174555 | AK031620.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174555 | NM_001098810.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104348 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167485 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09724546 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09724554 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27990329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27990332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09346277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09386388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09724554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09110457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09535551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09114080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27990390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09110449 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10379985 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379986 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379983 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455894_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451845_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10379981 | - | - |
| Mouse Genome 430 2.0 Array | 1451845_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451845_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1455894_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455894_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217057_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217057_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100012365.mm.1 | NM_175004.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012365.mm.1 | NM_175004.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012365.mm.1 | TC1100001321.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100012373.mm.1 | NM_001098810.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012373.mm.1 | NM_001098810.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012373.mm.1 | NM_175004.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012373.mm.1 | NM_175004.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012373.mm.1 | TC1100001321.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100012372.mm.1 | NM_001098810.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012372.mm.1 | NM_001098810.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012372.mm.1 | NM_175004.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012372.mm.1 | NM_175004.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012372.mm.1 | TC1100001321.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100012374.mm.1 | NM_001098810.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012374.mm.1 | NM_001098810.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012374.mm.1 | TC1100001321.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100012370.mm.1 | NM_175004.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012370.mm.1 | NM_175004.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012370.mm.1 | TC1100001321.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100012367.mm.1 | NM_001098810.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012367.mm.1 | NM_001098810.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012367.mm.1 | NM_175004.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012367.mm.1 | NM_175004.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012367.mm.1 | TC1100001321.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451845_a_at | - | - |
| Mouse Expression Set 430 | 1455894_PM_at | - | - |
| Mouse Expression Set 430 | 1451845_PM_a_at | - | - |
| Mouse Expression Set 430 | 1455894_at | - | - |
| Murine Genome U74v2 Set | 160719_at | - | - |
| Murine Genome U74v2 Set | 160719_PM_at | - | - |
| Murine Genome U74v2 Set | 171216_PM_r_at | - | - |
| Murine Genome U74v2 Set | 171216_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451845_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451845_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455894_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455894_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451845_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455894_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451845_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455894_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 912338 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106380138 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 770112 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 730270 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106380138 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000770112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000730270 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233095 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2633993 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233095 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238213 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2633993 | - | - |
| mouse-6 v1.1 (Illumina) | GI_33239414-S | - | - |
| mouse-6 v1.1 (Illumina) | scl41093.3_55-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001497.1_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106380138 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 770112 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 730270 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41093.3_55-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001497.1_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770112 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106380138 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 730270 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1238213 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2633993 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238213 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2633993 | - | - |

Ptrh2
Genomic Map