Assay Details
| Gene Symbol: Mrpl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026591.3 | NP_080867.2 | 5 | 680 | 157 | |
| GenBank mRNA | AK003362.1 | - | 5 | 684 | 157 | |
| AK008117.1 | - | 5 | 650 | 157 | ||
| AK008961.1 | - | 5 | 651 | 157 | ||
| AK013394.1 | - | 5 | 624 | 157 | ||
| AK016190.1 | - | 5 | 678 | 157 | ||
| AK019976.1 | - | 5 | 681 | 157 | ||
| AK020157.1 | - | 5 | 681 | 157 | ||
| AK167623.1 | - | 5 | 842 | 157 | ||
| BC004736.1 | - | 4 | 594 | 3498625 | 157 | |
| BC025506.1 | - | 5 | 693 | 5251655 | 157 | |
| BC031730.1 | - | 5 | 607 | 3991689 | 157 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22310708 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22310706 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22310703 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114288 | BC094052.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114288 | BC012224.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114288 | AK082223.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114288 | AK035389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2114288 | NM_153562.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451985 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 752980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 866582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133335 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149387 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114288 | BC094052.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114288 | BC012224.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114288 | AK082223.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114288 | AK035389.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2114288 | NM_153562.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22310720 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04979436 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149387 | - | - |
| Whole Mouse Genome G4122A | A_51_P203597 | AK020157.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127666 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149387 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03028323 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03028325 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03028312 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03028311 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03028319 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03028322 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22310708 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22310703 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22310706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22310696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03028322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02957782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02402357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03028324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03028311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03088397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02605263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22310693 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493100 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428709_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493094 | - | - |
| Mouse Genome 430 2.0 Array | 1428709_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1428709_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67707_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67707_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300004773.mm.1 | TC0300000708.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004778.mm.1 | NM_026591.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004778.mm.1 | NM_026591.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004778.mm.1 | TC0300000708.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004781.mm.1 | TC0300000708.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004777.mm.1 | NM_026591.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004777.mm.1 | NM_026591.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004777.mm.1 | TC0300000708.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004779.mm.1 | NM_026591.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004779.mm.1 | NM_026591.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004779.mm.1 | TC0300000708.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428709_PM_a_at | - | - |
| Mouse Expression Set 430 | 1428709_a_at | - | - |
| Murine Genome U74v2 Set | 101067_at | - | - |
| Murine Genome U74v2 Set | 101067_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428709_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428709_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428709_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428709_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1170014 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001170014 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2724536 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2724536 | - | - |
| mouse-6 v1.1 (Illumina) | scl23015.4.5_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1170014 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23015.4.5_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1170014 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2802601 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2802601 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl23015.4.5_22-S | - | - |

Mrpl24
Genomic Map