Assay Details
| Gene Symbol: 1700047M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015458.1 | 2 | 482 | 138 | ||
| GenBank mRNA | AK006718.1 | - | 2 | 266 | 138 | |
| AK019698.1 | - | 2 | 261 | 138 | ||
| AK047804.1 | - | 2243 | 138 | |||
| BC048591.1 | - | 2 | 482 | 138 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | degenerative spermatocyte homolog 1 (Drosophila) | Degs1 |
AA536663, Degs, Des1, Mdes |
-20.2kb | Anti-sense | Chr. 1 - 182275770 - 182282759 on NCBI Build GRCm38 |
| Downstream | F-box protein 28 | Fbxo28 |
4833428J17Rik, 5730505P19Rik, D1Ertd578e, Fbx28, mKIAA0483 |
9.9kb | Anti-sense | Chr. 1 - 182313102 - 182341606 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700047M11Rik
Genomic Map