Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | split hand/foot malformation (ectrodactyly) type 1 | Shfm1 |
DSS1, Shfdg1, Shfg |
-276.9kb | Sense | Chr. 6 - 6558275 - 6578658 on NCBI Build GRCm38 |
| Downstream | distal-less homeobox 6 | Dlx6 | 7.1kb | Anti-sense | Chr. 6 - 6863334 - 6867970 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map