Assay Details
| Gene Symbol: Gm16063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046178.1 | 4 | 1534 | 134 | ||
| GenBank mRNA | AK090161.1 | - | 4 | 1534 | 134 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
T-box 3, opposite strand 1 |
Tbx3os1 | Gm2301 | Anti-sense | Chr. 5 - 119597977 - 119671331 on NCBI Build GRCm38 | |
| Downstream | T-box 3 | Tbx3 | D5Ertd189e | 35.7kb | Sense | Chr. 5 - 119670160 - 119684724 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay targets the same location as MTA transcript cluster(s) TC0500001325.mm
This assay targets the same location as RTA transcript cluster(s) TC1200001662.rn
This assay targets the same location as RTA transcript cluster(s) TC1200001662.rn
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 258790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858172 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124996 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04675138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04227647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04233173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04268396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04806358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04883316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04258530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04178361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04738580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04863565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04705718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04910508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04258533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04253078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24033688 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0500011542.mm.1 | NR_046178.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011542.mm.1 | NR_046178.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011542.mm.1 | TC0500001325.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011544.mm.1 | NR_046178.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011544.mm.1 | NR_046178.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011544.mm.1 | TC0500001325.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011541.mm.1 | NR_046178.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011541.mm.1 | NR_046178.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011541.mm.1 | TC0500001325.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011543.mm.1 | NR_046178.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011543.mm.1 | NR_046178.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011543.mm.1 | TC0500001325.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 420637 | - | - |

Gm16063
Genomic Map