Assay Details
| Gene Symbol: Btg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297747.1 | NP_001284676.1 | 4 | 1073 | 79 | |
| NM_009770.3 | NP_033900.1 | 5 | 1269 | 79 | ||
| GenBank mRNA | AK144415.1 | - | 5 | 1220 | 79 | |
| BC012705.1 | - | 5 | 1214 | 3980599 | 79 | |
| BC094027.1 | - | 5 | 1230 | 4457150 | 79 | |
| D83745.1 | - | 5 | 1239 | 79 | ||
| Z72000.1 | - | 5 | 1250 | 79 | ||
| Gene Symbol: Gm7334 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002700.1 | 1 | 1218 | 79 | ||
| GenBank mRNA | AA388647.1 | - | 1 | 315 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895867 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895861 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895871 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895869 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895870 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895862 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895866 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895865 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895864 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895863 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972322 | NR_002700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972322 | AA388647.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972322 | Z72000.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972322 | BC094027.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972322 | BC012705.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549001 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 463734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742296 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635838 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507535 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736313 | AK030546.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736313 | AK049948.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736313 | NM_178194.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93160 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135051 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114251 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164001 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130529 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972322 | NR_002700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972322 | AA388647.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972322 | Z72000.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972322 | BC094027.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972322 | BC012705.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340601 | NR_002700.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340601 | AA265715.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340601 | Z72000.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340601 | BC094027.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P340601 | BC147656.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30895883 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130529 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114251 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08654730 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164001 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08654716 | - | - |
| Whole Mouse Genome G4122A | A_51_P340601 | NR_002700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340601 | AA265715.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340601 | Z72000.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340601 | BC094027.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P340601 | BC147656.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139045 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08654716 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114251 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164001 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130529 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13007606 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13007613 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895866 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895867 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895864 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895861 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895865 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895870 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895869 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895863 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895862 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30895871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30895855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13007605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12990280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12966820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12751594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12966817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12966818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12913206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12825523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13007610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30895782 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440422 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10440419 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445875 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10440419 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445875 | - | - |
| Mouse Genome 430 2.0 Array | 1449007_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449007_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600015070.mm.1 | NM_009770.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015070.mm.1 | NR_002700.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015070.mm.1 | TC1600001939.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449007_PM_at | - | - |
| Mouse Expression Set 430 | 1449007_at | - | - |
| Mouse Expression Set 430 | 1449007_PM_at | - | - |
| Mouse Expression Set 430 | 1449007_at | - | - |
| Murine Genome U74v2 Set | 96146_at | - | - |
| Murine Genome U74v2 Set | 96146_at | - | - |
| Murine Genome U74v2 Set | 96146_PM_at | - | - |
| Murine Genome U74v2 Set | 96146_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449007_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449007_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449007_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449007_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 897083 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 7050079 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007050079 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2685011 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2685011 | - | - |
| mouse-6 v1.1 (Illumina) | scl012228.1_44-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7050079 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl012228.1_44-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7050079 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3117483 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2945956 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3042627 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3117483 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2945956 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3042627 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2685011 | - | - |

Btg3
Genomic Map