Assay Details
| Gene Symbol: Gm13583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC023004.1 | - | 3 | 1361 | 124 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 13580 | Gm13580 | OTTMUSG00000012842 | -140.2kb | Sense | Chr. 2 - 60411525 - 60412574 on NCBI Build GRCm38 |
| Downstream | integrin beta 6 | Itgb6 |
2210409C20Rik, 4831415H04Rik |
44.4kb | Anti-sense | Chr. 2 - 60598292 - 60722603 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm13583
Genomic Map