Assay Details
| Gene Symbol: Aldh1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027406.1 | NP_081682.1 | 4 - 5 | 680 | 77 | |
| GenBank mRNA | BC022112.1 | - | 4 - 5 | 680 | 77 | |
| BC024055.1 | - | 4 - 5 | 637 | 5098789 | 77 | |
| BC025939.1 | - | 4 - 5 | 662 | 5099360 | 77 | |
| BC028817.1 | - | 4 - 5 | 627 | 5101815 | 77 | |
| BC030722.1 | - | 4 - 5 | 664 | 5098671 | 77 | |
| BC030723.1 | - | 4 - 5 | 596 | 5100007 | 77 | |
| BC030727.1 | - | 4 - 5 | 666 | 5099026 | 77 | |
| BC030730.1 | - | 4 - 5 | 680 | 5101835 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh1l1
Genomic Map