Assay Details
| Gene Symbol: Gm4221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079064.1 | - | 2 | 2926 | 119 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 608 | Zfp608 |
4932417D18Rik, D430007A19Rik, Znf608 |
-83.9kb | Anti-sense | Chr. 18 - 54888045 - 54992598 on NCBI Build GRCm38 |
| Downstream | GRAM domain containing 3 | Gramd3 |
9030613F08Rik, 9130427A09Rik, AW061306, NS3TP2 |
1.3mb | Sense | Chr. 18 - 56387485 - 56503792 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm4221
Genomic Map