Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 40811 |
Gm40811 | -21.8kb | Anti-sense | Chr. 10 - 117145168 - 117149728 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930423D24 gene | 4930423D24Rik | 5.4kb | Anti-sense | Chr. 10 - 117177625 - 117181110 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map