Assay Details
| Gene Symbol: Nova2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029877.3 | NP_001025048.2 | 4 | 88 | ||
| GenBank mRNA | BC062110.1 | - | 1 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 61 | Ccdc61 |
C530028I08Rik, Gm159 |
-47.1kb | Anti-sense | Chr. 7 - 18890883 - 18910412 on NCBI Build GRCm38 |
| Downstream | nanos homolog 2 (Drosophila) | Nanos2 | nos2 | 22.2kb | Sense | Chr. 7 - 18987524 - 18988962 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Nova2
Genomic Map