Assay Details
| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK165779.1 | - | 1 | 2262 | 77 | |
| BC058257.1 | - | 1 | 2234 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ckb
Genomic Map