Assay Details
| Gene Symbol: Zfp251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007568.2 | NP_001007569.1 | 3 - 4 | 1027 | 134 | |
| GenBank mRNA | AK047148.1 | - | 2 - 3 | 1248 | 134 | |
| AK140082.1 | - | 3 - 4 | 1018 | 134 | ||
| AK153967.1 | - | 2 - 3 | 868 | 134 | ||
| BC059071.1 | - | 3 - 4 | 1027 | 6826152 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350969 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350974 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350972 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350970 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350973 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P15340 | BC041112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P15340 | BC059071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P15340 | AK140082.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P15340 | AK153967.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P15340 | AK047299.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337590 | AF071316.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337590 | AK010417.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337590 | AK010726.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337590 | AK013155.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337590 | AK019122.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337590 | BC003724.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337590 | NM_012003.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872851 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166498 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 442005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345838 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868929 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 463235 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346504 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94289 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165525 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179316 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P15340 | BC041112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P15340 | BC059071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P15340 | AK140082.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P15340 | AK153967.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P15340 | AK047299.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30351001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350978 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179316 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08423545 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08423562 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165525 | - | - |
| Whole Mouse Genome G4122A | A_52_P314940 | BC041112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P314940 | BC059071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P314940 | AK140082.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P314940 | AK153967.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P314940 | AK047148.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P564005 | BC041112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P564005 | AK047299.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P564005 | AK018584.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165525 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179316 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12186364 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11939487 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11908141 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11908138 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12016993 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12186359 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12372067 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350969 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350973 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350972 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350970 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30351001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30351011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30351009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30351005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30351003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12406548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11919214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11881275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12135838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12327391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11916363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11970783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12291991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12261607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11939501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11939489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12327393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11939495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12135841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12414365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12327390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11919211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12416484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11939480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12343944 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430131 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430128 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430130 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430129 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453034_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430127 | - | - |
| Mouse Genome 430 2.0 Array | 1453034_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453034_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500013948.mm.1 | NM_001007568.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013948.mm.1 | NM_001007568.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013948.mm.1 | TC1500001793.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500013949.mm.1 | NM_001007568.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013949.mm.1 | NM_001007568.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013949.mm.1 | TC1500001793.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500013947.mm.1 | NM_001007568.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013947.mm.1 | NM_001007568.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013947.mm.1 | TC1500001793.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453034_at | - | - |
| Mouse Expression Set 430 | 1453034_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453034_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453034_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2524263 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2870623 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2870623 | - | - |

Zfp251
Genomic Map