Assay Details
| Gene Symbol: Plp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019755.4 | NP_062729.1 | 3 - 4 | 387 | 149 | |
| GenBank mRNA | AB031292.1 | - | 3 - 4 | 405 | 149 | |
| AK003522.1 | - | 3 - 4 | 442 | 149 | ||
| AK011282.1 | - | 3 - 4 | 441 | 149 | ||
| AK154228.1 | - | 3 - 4 | 451 | 149 | ||
| BC083078.1 | - | 3 - 4 | 430 | 6477560 | 149 | |
| BC092095.1 | - | 3 - 4 | 406 | 3989985 | 149 | |
| BC119088.1 | - | 3 - 4 | 387 | 8733837 | 149 | |
| BC119090.1 | - | 3 - 4 | 387 | 8733839 | 149 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Plp2
Genomic Map