Assay Details
| Gene Symbol: Plp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019755.4 | NP_062729.1 | 4 - 5 | 475 | 116 | |
| GenBank mRNA | AB031292.1 | - | 4 - 5 | 493 | 116 | |
| AK003522.1 | - | 4 - 5 | 530 | 116 | ||
| AK011282.1 | - | 4 - 5 | 529 | 116 | ||
| AK012816.1 | - | 3 - 4 | 662 | 116 | ||
| AK154228.1 | - | 4 - 5 | 539 | 116 | ||
| BC083078.1 | - | 4 - 5 | 518 | 6477560 | 116 | |
| BC092095.1 | - | 4 - 5 | 494 | 3989985 | 116 | |
| BC119088.1 | - | 4 - 5 | 475 | 8733837 | 116 | |
| BC119090.1 | - | 4 - 5 | 475 | 8733839 | 116 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239592 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239595 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239594 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P418644 | BC119088.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P418644 | BC119090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P418644 | AK003522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P418644 | AK011282.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P418644 | AK012816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P418644 | BC119088.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P418644 | BC119090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P418644 | AK003522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P418644 | AK011282.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P418644 | AK012816.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 24287 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179444 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P418644 | BC119088.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P418644 | BC119090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P418644 | AK003522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P418644 | AK011282.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P418644 | AK012816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P418644 | BC119088.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P418644 | BC119090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P418644 | AK003522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P418644 | AK011282.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P418644 | AK012816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2090194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239597 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179444 | - | - |
| Whole Mouse Genome G4122A | A_51_P243532 | BC092095.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243532 | BC119088.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243532 | BC119090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243532 | AK003522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243532 | AK011282.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418644 | BC119088.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418644 | BC119090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418644 | AK003522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418644 | AK011282.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418644 | AK012816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418644 | BC119088.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418644 | BC119090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418644 | AK003522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418644 | AK011282.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P418644 | AK012816.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108334 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179444 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15050520 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15209138 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15209140 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15180793 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239594 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239595 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15050516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15209144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15050519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15209140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239595 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10603347 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473023 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10603348 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10603352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10603351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10603353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10603350 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411362 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411361 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10603349 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411360 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453572_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456247_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10473022 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10411359 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10603346 | - | - |
| Mouse Genome 430 2.0 Array | 1453572_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1453572_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456247_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1456247_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00011232.mm.1 | TC0X00001780.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00011233.mm.1 | NM_019755.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00011233.mm.1 | TC0X00001780.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456247_PM_x_at | - | - |
| Mouse Expression Set 430 | 1456247_x_at | - | - |
| Mouse Expression Set 430 | 1453572_PM_a_at | - | - |
| Mouse Expression Set 430 | 1453572_a_at | - | - |
| Murine Genome U74v2 Set | 93323_at | - | - |
| Murine Genome U74v2 Set | 93323_PM_at | - | - |
| Murine Genome U74v2 Set | 171551_PM_i_at | - | - |
| Murine Genome U74v2 Set | 171551_i_at | - | - |
| Murine Genome U74v2 Set | 140736_PM_at | - | - |
| Murine Genome U74v2 Set | 140736_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453572_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456247_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453572_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456247_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456247_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456247_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453572_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453572_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 915102 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2100167 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002100167 | - | - |
| mouse-6 v1.1 (Illumina) | scl018824.1_278-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2100167 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl018824.1_278-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100167 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3001616 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2602858 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3001616 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl018824.1_278-S | - | - |

Plp2
Genomic Map