Assay Details
| Gene Symbol: Flad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC009152.1 | - | 1251 | 165 | ||
| Gene Symbol: Lenep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC006806.1 | - | 459 | 3152957 | 165 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327232 | AK044501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327232 | NM_020517.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327232 | AK053703.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327232 | BC006806.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327232 | CK627663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327232 | AK044501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327232 | NM_020517.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327232 | AK053703.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327232 | BC006806.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327232 | CK627663.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369745 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327232 | AK044501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327232 | NM_020517.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327232 | AK053703.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327232 | BC006806.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327232 | CK627663.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327232 | AK044501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327232 | NM_020517.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327232 | AK053703.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327232 | BC006806.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327232 | CK627663.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327232 | AK044501.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327232 | NM_020517.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327232 | AK053703.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327232 | BC006806.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327232 | CK627663.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327232 | AK044501.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327232 | NM_020517.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327232 | AK053703.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327232 | BC006806.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327232 | CK627663.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319168 | - | - |
| Whole Mouse Genome G4122A | A_52_P101551 | EL956268.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P101551 | NM_020517.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P101551 | AK053703.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P101551 | BC006806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P101551 | CK627663.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327232 | AK044501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327232 | NM_020517.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327232 | AK053703.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327232 | BC006806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327232 | CK627663.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327232 | AK044501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327232 | NM_020517.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327232 | AK053703.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327232 | BC006806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327232 | CK627663.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138824 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107757 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02904690 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02904686 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03169529 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03169546 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03169535 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03169532 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02904683 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03169541 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02904692 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03169540 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02904691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03101585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02435149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02422774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02904690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03169529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03101588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02422767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02422781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03101582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03140644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03169526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03169540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03101589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03101579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02904692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02904680 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499621 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499623 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499622 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499625 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422309_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499620 | - | - |
| Mouse Genome 430 2.0 Array | 1424421_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424423_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424423_at | - | - |
| Mouse Genome 430 2.0 Array | 1424421_at | - | - |
| Mouse Genome 430 2.0 Array | 1422309_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424422_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1424422_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422309_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57275_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57275_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300018118.mm.1 | NM_177041.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018118.mm.1 | NM_177041.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018118.mm.1 | TC0300002351.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300018111.mm.1 | TC0300002351.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300018114.mm.1 | TC0300002351.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424423_at | - | - |
| Mouse Expression Set 430 | 1424421_at | - | - |
| Mouse Expression Set 430 | 1424423_PM_at | - | - |
| Mouse Expression Set 430 | 1424421_PM_at | - | - |
| Mouse Expression Set 430 | 1424422_s_at | - | - |
| Mouse Expression Set 430 | 1422309_a_at | - | - |
| Mouse Expression Set 430 | 1424422_PM_s_at | - | - |
| Mouse Expression Set 430 | 1422309_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422309_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424422_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422309_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424423_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424421_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424423_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424422_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424421_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422309_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422309_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4850369 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850369 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252499 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2670942 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2670942 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252499 | - | - |
| mouse-6 v1.1 (Illumina) | scl057275.1_158-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850369 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl057275.1_158-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850369 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1235612 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235612 | - | - |

Flad1
Genomic Map