Assay Details
| Gene Symbol: Klhl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172871.2 | NP_766459.2 | 1 | 2868 | 153 | |
| GenBank mRNA | AK049803.1 | - | 1 | 2824 | 153 | |
| AK129338.1 | - | 1 | 2780 | 153 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Klhl9
Genomic Map