Assay Details
| Gene Symbol: Rnase12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011875.2 | NP_001011875.1 | 1 | 404 | 73 | |
| GenBank mRNA | BC146014.1 | - | 1 | 404 | 40131317 | 73 |
| BC146016.1 | - | 1 | 404 | 40131319 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109407 | NM_001011875.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109407 | BC146014.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109407 | BC146016.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2109407 | NM_001011875.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2109407 | BC146014.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2109407 | BC146016.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29599838 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11647081 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11647071 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11647075 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11647069 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11647065 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11647077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11341946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11567730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11420971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11567734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11647069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11647076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11828626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11567727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11647081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11276732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11647065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11647072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11761656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11276738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11647077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29599827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11567719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11484635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11491809 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10419560 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419559 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400015027.mm.1 | NM_001011875.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015027.mm.1 | TC1400001997.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400015028.mm.1 | NM_001011875.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400015028.mm.1 | TC1400001997.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3162426 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3162426 | - | - |
| mouse-6 v1.1 (Illumina) | GI_59858540-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3162426 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3162426 | - | - |

Rnase12
Genomic Map