Assay Details
| Gene Symbol: Crybb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020891.1 | - | 4 - 5 | 359 | 158 | |
| BC085185.1 | - | 5 - 6 | 501 | 30679384 | 158 | |
| M60559.1 | - | 3 - 4 | 326 | 158 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crybb2
Genomic Map