Assay Details
| Gene Symbol: Pou4f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011143.4 | NP_035273.3 | 1 - 2 | 387 | 127 | |
| GenBank mRNA | AY706205.1 | - | 1 - 2 | 387 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pou4f1
Genomic Map