Assay Details
| Gene Symbol: Dbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005232.1 | NP_001005232.1 | 2 - 3 | 589 | 180 | |
| GenBank mRNA | BC082541.1 | - | 2 - 3 | 589 | 5718470 | 180 |
| S75837.1 | - | 2 - 3 | 602 | 180 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dbx1
Genomic Map