Assay Details
| Gene Symbol: Rtp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008230.3 | NP_001008231.1 | 2 | 2044 | 154 | |
| GenBank mRNA | BC049158.1 | - | 2 | 2044 | 6390582 | 154 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30605570 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155136 | NM_001008230.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155136 | BC049158.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768958 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72186 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198806 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13318 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149466 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155136 | NM_001008230.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155136 | BC049158.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30605610 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149466 | - | - |
| Whole Mouse Genome G4122A | A_52_P216820 | NM_001008230.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P216820 | BC049158.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149466 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12834177 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12834195 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12834187 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12834189 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30605570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30605578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12546288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13054013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12584909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12507758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12834195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12479140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12739474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12834178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12834187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12834164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12507765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12834183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13050127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13050128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12834163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13050130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12887262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30605578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12939280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12802621 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10438735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438737 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10438733 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600011632.mm.1 | NM_001008230.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600011632.mm.1 | NM_001008230.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600011632.mm.1 | TC1600001399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600011633.mm.1 | NM_001008230.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600011633.mm.1 | NM_001008230.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600011633.mm.1 | TC1600001399.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 135281_PM_at | - | - |
| Murine Genome U74v2 Set | 135281_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 540052 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2530023 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2855019 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2855019 | - | - |

Rtp2
Genomic Map