Assay Details
| Gene Symbol: Mpzl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK041037.1 | - | 2 | 3209 | 144 | |
| AK049878.1 | - | 2 | 3207 | 144 | ||
| AK155483.1 | - | 2 | 3144 | 144 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mpzl3
Genomic Map