Assay Details
| Gene Symbol: Rps10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025963.3 | NP_080239.1 | 5 - 6 | 499 | 87 | |
| GenBank mRNA | AK002558.1 | - | 5 - 6 | 497 | 87 | |
| AK008797.1 | - | 5 - 6 | 500 | 87 | ||
| AK011065.1 | - | 5 - 6 | 492 | 87 | ||
| AK020219.1 | - | 5 - 6 | 500 | 87 | ||
| AK145441.1 | - | 5 - 6 | 492 | 87 | ||
| AK150803.1 | - | 5 - 6 | 498 | 87 | ||
| AK157104.1 | - | 5 - 6 | 498 | 87 | ||
| BC003853.1 | - | 5 - 6 | 486 | 3601013 | 87 | |
| BC019725.1 | - | 5 - 6 | 486 | 3497741 | 87 | |
| BC086919.1 | - | 5 - 6 | 498 | 6763104 | 87 | |
| BC089323.1 | - | 4 - 5 | 475 | 6814929 | 87 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117589 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117613 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117590 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117612 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117592 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117614 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117588 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117591 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051047 | BC003853.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051047 | BC019725.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051047 | BC086919.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051047 | BC089323.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051047 | NM_025963.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183604 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970682 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208004 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165007 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145583 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P140711 | BC003853.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P140711 | BC086919.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051047 | BC003853.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051047 | BC019725.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051047 | BC086919.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051047 | BC089323.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051047 | NM_025963.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P169156 | BC003853.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P169156 | BC019725.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P169156 | BC086919.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P169156 | BC089323.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P169156 | NM_025963.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31117593 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145583 | - | - |
| Whole Mouse Genome G4122A | A_51_P169156 | BC003853.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P169156 | BC019725.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P169156 | BC086919.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P169156 | BC089323.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P169156 | NM_025963.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129475 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140711 | BC003853.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140711 | BC086919.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124694 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145583 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13585168 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13585171 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13585163 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13446363 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13516620 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13585174 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13585166 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117614 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117612 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117590 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117592 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117591 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117613 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117588 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31117589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31117599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13535350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13552466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13515229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13342675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13365509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13272266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13428644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13515230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31117610 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449371 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494297 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494298 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449374 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438566 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449375 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438565 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449376 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449372 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434854_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436384_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439374_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438723_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456497_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416719_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494296 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449370 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10438564 | - | - |
| Mouse Genome 430 2.0 Array | 1439374_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436384_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434854_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1434854_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1436384_at | - | - |
| Mouse Genome 430 2.0 Array | 1456497_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1439374_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438723_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456497_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1416719_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1438723_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416719_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700016009.mm.1 | NM_025963.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016009.mm.1 | NM_025963.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016009.mm.1 | TC1700001742.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700016010.mm.1 | NM_025963.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016010.mm.1 | NM_025963.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016010.mm.1 | TC1700001742.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416719_PM_a_at | - | - |
| Mouse Expression Set 430 | 1439374_PM_x_at | - | - |
| Mouse Expression Set 430 | 1439374_x_at | - | - |
| Mouse Expression Set 430 | 1456497_x_at | - | - |
| Mouse Expression Set 430 | 1434854_a_at | - | - |
| Mouse Expression Set 430 | 1434854_PM_a_at | - | - |
| Mouse Expression Set 430 | 1456497_PM_x_at | - | - |
| Mouse Expression Set 430 | 1436384_PM_at | - | - |
| Mouse Expression Set 430 | 1416719_a_at | - | - |
| Mouse Expression Set 430 | 1438723_a_at | - | - |
| Mouse Expression Set 430 | 1438723_PM_a_at | - | - |
| Mouse Expression Set 430 | 1436384_at | - | - |
| Murine Genome U74v2 Set | 99093_PM_at | - | - |
| Murine Genome U74v2 Set | 99093_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436384_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416719_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416719_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438723_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456497_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434854_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436384_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439374_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438723_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456497_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439374_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434854_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456497_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416719_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456497_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439374_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438723_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439374_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436384_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436384_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434854_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434854_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416719_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438723_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 894633 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4560673 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4560673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004560673 | - | - |
| mouse-6 v1.1 (Illumina) | scl067097.5_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4560673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067097.5_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4560673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2897617 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2897617 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067097.5_20-S | - | - |

Rps10
Genomic Map