Assay Details
| Gene Symbol: Wnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175638.3 | NP_783569.1 | 16 - 17 | 3470 | 92 | |
| GenBank mRNA | AK156847.1 | - | 3 - 4 | 835 | 92 | |
| AY184228.1 | - | 16 - 17 | 3439 | 92 | ||
| AY187027.1 | - | 16 - 17 | 3395 | 92 | ||
| BC026591.1 | - | 7 - 8 | 1409 | 92 | ||
| BC026679.1 | - | 9 - 10 | 1669 | 92 | ||
| BC037114.1 | - | 14 - 15 | 2794 | 92 | ||
| BC043677.1 | - | 16 - 17 | 2843 | 92 | ||
| BC096453.1 | - | 16 - 17 | 3426 | 4234311 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wnk4
Genomic Map