Assay Details
| Gene Symbol: Nanos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194064.2 | NP_918953.2 | 1 | 1348 | 83 | |
| GenBank mRNA | AB095972.1 | - | 1 | 1349 | 83 | |
| JQ936992.1 | - | 1 | 1278 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mRNA catabolic process
regulation of translation
multicellular organism development
spermatogenesis
negative regulation of translation
cell differentiation
germ-line stem cell population maintenance
cell quiescence
negative regulation of meiotic nuclear division
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
RNA binding
mRNA binding
protein binding
zinc ion binding
metal ion binding
mRNA catabolic process
regulation of translation
multicellular organism development
spermatogenesis
negative regulation of translation
cell differentiation
germ-line stem cell population maintenance
cell quiescence
negative regulation of meiotic nuclear division
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
RNA binding
mRNA binding
protein binding
zinc ion binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096071 | NM_194064.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096071 | AB095972.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649303 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140051 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096071 | NM_194064.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096071 | AB095972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P624495 | NM_194064.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P624495 | AB095972.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24990761 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140051 | - | - |
| Whole Mouse Genome G4122A | A_52_P624495 | NM_194064.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P624495 | AB095972.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105554 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140051 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06089487 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06216590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24990738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06557380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06651708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06656620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06469140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06089490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24990757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06216590 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10550520 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10550519 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6290168 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006290168 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2637484 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2637484 | - | - |
| mouse-6 v1.1 (Illumina) | scl0378430.1_272-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6290168 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0378430.1_272-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6290168 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2637484 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2637484 | - | - |

Nanos2
Genomic Map