Assay Details
| Gene Symbol: Gmnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013761.2 | NP_001013783.1 | 4 - 5 | 575 | 93 | |
| NM_001285916.1 | NP_001272845.1 | 3 - 4 | 669 | 93 | ||
| NM_001285918.1 | NP_001272847.1 | 3 - 4 | 400 | 93 | ||
| GenBank mRNA | AK141059.1 | - | 3 - 4 | 669 | 93 | |
| AK160719.1 | - | 3 - 4 | 369 | 93 | ||
| BC086669.1 | - | 3 - 4 | 330 | 30639759 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmnc
Genomic Map