Assay Details
| Gene Symbol: Puf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164600.1 | NP_001158072.1 | 6 - 7 | 607 | 103 | |
| NM_028364.2 | NP_082640.2 | 7 - 8 | 688 | 103 | ||
| NM_133691.5 | NP_598452.2 | 6 - 7 | 637 | 103 | ||
| GenBank mRNA | AK012434.1 | - | 6 - 7 | 577 | 103 | |
| AK075960.1 | - | 6 - 7 | 605 | 103 | ||
| AK146534.1 | - | 7 - 8 | 659 | 103 | ||
| AK149632.1 | - | 7 - 8 | 654 | 103 | ||
| AK151526.1 | - | 6 - 7 | 601 | 103 | ||
| AK168789.1 | - | 6 - 7 | 604 | 103 | ||
| BC010601.1 | - | 6 - 7 | 607 | 3257153 | 103 | |
| BC119437.1 | - | 4 - 5 | 344 | 103 | ||
| BC148274.1 | - | 4 - 5 | 344 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Puf60
Genomic Map