Assay Details
| Gene Symbol: Lemd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC048759.1 | - | 3 | 460 | 69 | |
| Gene Symbol: Gm10188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015739.1 | - | 1 | 69 | ||
More Information
Set Membership: |
|

Lemd1
Genomic Map