Assay Details
| Gene Symbol: Gas2l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033331.2 | NP_001028503.2 | 9 - 10 | 969 | 118 | |
| NM_001079876.1 | NP_001073345.1 | 9 - 10 | 972 | 118 | ||
| NM_001284344.1 | NP_001271273.1 | 9 - 10 | 969 | 118 | ||
| GenBank mRNA | AK136054.1 | - | 9 - 10 | 982 | 118 | |
| AK164505.1 | - | 9 - 10 | 958 | 118 | ||
| AK170634.1 | - | 9 - 10 | 904 | 118 | ||
| AK170750.1 | - | 9 - 10 | 894 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gas2l3
Genomic Map