| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700004142.mm.1 |
NM_011287.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004142.mm.1 |
NM_011287.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004142.mm.1 |
TC1700000477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004149.mm.1 |
NM_011287.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004149.mm.1 |
NM_011287.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004149.mm.1 |
TC1700000477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004155.mm.1 |
NM_011287.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004155.mm.1 |
NM_011287.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004155.mm.1 |
TC1700000477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004158.mm.1 |
NM_011287.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004158.mm.1 |
NM_011287.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004158.mm.1 |
TC1700000477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004150.mm.1 |
NM_011287.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004150.mm.1 |
NM_011287.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004150.mm.1 |
TC1700000477.mm.1 |
MTA TC |