Assay Details
| Gene Symbol: Cd200r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206535.1 | NP_996258.1 | 2 - 3 | 278 | 80 | |
| GenBank mRNA | AY230198.1 | - | 2 - 3 | 278 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099039 | NM_206535.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099039 | AY230198.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641948 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338947 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475878 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891447 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861687 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675823 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 840019 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539044 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338376 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95077 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104967 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133751 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137691 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148721 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099039 | NM_206535.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099039 | AY230198.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30723867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30723856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30723859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30723863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30723872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30723871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30723861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30723860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30723865 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08584324 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148721 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03184934 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08584323 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03184944 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137691 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108307 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137691 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148721 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12688419 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12688426 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12688428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30723860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12894824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12688426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12688428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12688419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30723874 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10435933 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435936 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435932 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10435931 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10435930 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600005836.mm.1 | NM_206535.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005836.mm.1 | NM_206535.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005836.mm.1 | TC1600000665.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600005834.mm.1 | NM_206535.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005834.mm.1 | NM_206535.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005834.mm.1 | TC1600000665.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600005838.mm.1 | NM_206535.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005838.mm.1 | NM_206535.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005838.mm.1 | TC1600000665.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600005832.mm.1 | NM_206535.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005832.mm.1 | NM_206535.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005832.mm.1 | TC1600000665.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600005833.mm.1 | NM_206535.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005833.mm.1 | NM_206535.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005833.mm.1 | TC1600000665.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600005835.mm.1 | NM_206535.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005835.mm.1 | NM_206535.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005835.mm.1 | TC1600000665.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600005837.mm.1 | NM_206535.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005837.mm.1 | NM_206535.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005837.mm.1 | TC1600000665.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435313_at | - | - |
| Mouse Expression Set 430 | 1435313_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 705045 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3990021 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4610152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4610152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3990021 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004610152 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003990021 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239931 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231427 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231427 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239931 | - | - |
| mouse-6 v1.1 (Illumina) | scl0271375.3_22-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00271375.2_273-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4610152 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3990021 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00271375.2_273-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0271375.3_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3990021 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610152 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239931 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2688180 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2688182 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239931 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688182 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688180 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00271375.2_273-S | - | - |

Cd200r2
Genomic Map