Assay Details
| Gene Symbol: Ccr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166625.1 | NP_001160097.1 | 4 | 2230 | 153 | |
| NM_009913.6 | NP_034043.1 | 3 | 2162 | 153 | ||
| GenBank mRNA | AJ131357.2 | - | 4 | 2167 | 153 | |
| AK019478.1 | - | 2336 | 153 | |||
| AK050615.1 | - | 3 | 2163 | 153 | ||
| AK153975.1 | - | 3 | 2163 | 153 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccr9
Genomic Map